Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
Representative image only.
Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
Cole-Parmer | Item: 6893680
Replace mercury manometers for monitoring vacuum systems
₹111,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹111,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
6893680
₹111,000.00INR / Each
+ + + =
Total Price 253200.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • ±0.25% test gauge accuracy
  • 316 stainless steel wetted parts
  • Fast response—3 readings per second
  • 1 year warranty
More About this Item
Replace toxic mercury manomaters with these vacuum gauges! Use for monitoring laboratory or industrial vacuum manifolds, pumps, and systems. Large display provides better readability than mercury or liquid manometers. The 760 torr absolute scale measures atmospheric pressure when gauge port is open and reads zero at full vacuum.
Rugged epoxy-coated extruded aluminum case and rear cover, ABS/polycarbonate front bezel, front on/off push button, front accessible calibration adjustments.
1 Year Warranty
Specifications
  • Accuracy±0.25% full-scale ±1 digit
  • Wetted Materials316 SS
  • Casealuminum
  • Process Connection1/4" NPT(M)
  • Unit Of MeasurementTorr
  • Display3-digit LCD, 1/2" H
  • Battery Type115 VAC/12 VDC power supply (included)
  • Media CompatibilityGases compatible with wetted materials
  • Resolution1 torr
  • DescriptionDigital Gauge, 760 Torr; 1/4"; NPT(M)