Cole-Parmer Deuterium (D2) Detector Lamp for Interchim Puriflash PF215, PF430
Representative image only.
Cole-Parmer Deuterium (D2) Detector Lamp for Interchim Puriflash PF215, PF430
Cole-Parmer | Item: 1194121
Economical replacement deuterium lamps save you money
₹101,000.00INR / Each
₹101,000.00INR / Each
Product Description
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 1 year warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
1 Year Warranty
Specifications
  • Compatible WithInterchim PuriFlash (PF) 215, PF430, PF450, PF800, PF4100, PF1, SP2
  • DescriptionDeuterium (D2) Detector Lamp for Interchim Puriflash PF215, PF430