Cole-Parmer Deuterium (D2) Detector Lamp for Dionex / Gynkotek UVD 320, 340S, 160, 170S, 170U

Representative image only.

Cole-Parmer Deuterium (D2) Detector Lamp for Dionex / Gynkotek UVD 320, 340S, 160, 170S, 170U

Cole-Parmer | Item: 1194120

Economical replacement deuterium lamps save you money

₹62,900.00INR / Each

₹62,900.00INR / Each
Product Description

Key Features

  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Long life 2000 hour bulb
  • 1 year warranty

More About this Item

Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.

Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.

1 Year Warranty
Specifications
  • Compatible WithDionex/Gynkotek UVD160, 170S, 170U, 320, 340S, 340U
  • DescriptionDeuterium (D2) Detector Lamp for Dionex / Gynkotek UVD 320, 340S, 160, 170S, 170U
  • Warranty1 year
Reviews
Q&A