Cole-Parmer Deuterium (D2) Detector Lamp for Bischoff Lambda 1010
Representative image only.
Cole-Parmer Deuterium (D2) Detector Lamp for Bischoff Lambda 1010
Cole-Parmer | Item: 1194143
Economical replacement deuterium lamps save you money
₹127,000.00INR / Each
₹127,000.00INR / Each
Product Description
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 6 month warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
6 Month Warranty
Specifications
  • Compatible WithBischoff Lambda 1010
  • DescriptionDeuterium (D2) Detector Lamp for Bischoff Lambda 1010