Cole-Parmer Deuterium (D2) Detector Lamp for Biorad 206, 300, 1790, BioDimensions Microsampler 100
Representative image only.
Cole-Parmer Deuterium (D2) Detector Lamp for Biorad 206, 300, 1790, BioDimensions Microsampler 100
Cole-Parmer | Item: 1194140
Economical replacement deuterium lamps save you money
₹107,000.00INR / Each
₹107,000.00INR / Each
Product Description
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 6 month warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.
6 Month Warranty
Specifications
  • Compatible WithBiorad 206, 300, 1790; BioDimensions Microsampler 100
  • DescriptionDeuterium (D2) Detector Lamp for Biorad 206, 300, 1790, BioDimensions Microsampler 100