Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L

Representative image only.

Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L

Cole-Parmer | Item: 7890287

Automated dehumidification system offers convenience

₹292,000.00INR / Each

₹292,000.00INR / Each

Buy It Together

Current Item

Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L
Cole-Parmer DC-400 Automatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L
7890287

₹292,000.00INR / Each
+ +
Scienceware F42400-4001 Vacuum Desiccator Cabinet, 11L.
0890916

₹585,000.00INR / Each
=
Total Price 2137000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Two SS shelves and a key.

Key Features

  • Set relative humidity range (RH) between 25 and 55%
  • Built-in digital hygrometer allows easy monitoring of RH
  • View your samples through acrylic viewing panels
  • 2.47 cu ft (70 L) capacity
  • 3 year warranty

More About this Item

Keeping a constant RH level has never been easier. The electronic dehumidification system includes a permanent desiccant and a hygrometer to keep the relative humidity level between 25 and 55%. The desiccant automatically recycles by discharging absorbed moisture through an external shutter. Once it is recycled, desiccant is ready to absorb moisture again. Safely keep humidity sensitive products such as optical equipment, electronic parts, PCB boards, and reagents.

Aluminum frame and stainless steel (SS) shelves offer excellent durability. SS shelves have holes to ensure uniform humidity throughout chamber. A lock and key let you securely lock the cabinet to prevent unauthorized access.

The fuse used for acrylic automatic cabinet will stop operation if the temperature inside the cabinet exceeds 172°C.

3 Year Warranty
Specifications
  • Chamber Volume (cu ft)2.47
  • Chamber Volume (Liters)70
  • Interior Width (in)12.2
  • Interior Height (in)15.9
  • Interior Depth (in)18.8
  • Interior Width (cm)31
  • Interior Height (cm)40.5
  • Interior Depth (cm)47.8
  • Exterior Width (in)15
  • Exterior Height (InchShort)22.9
  • Exterior Depth (in)19.8
  • Exterior Width (cm)38
  • Exterior Height (cm)58.1
  • Exterior Depth (cm)50.2
  • MaterialAluminum frame, acrylic panels, and SS shelves
  • ColorClear
  • Power (VAC)110
  • Power (Hz)50/60
  • DescriptionAutomatic Desiccator Cabinet, 25 to 50% RH, Two Shelves; 70 L
  • Warranty2 Years
Reviews
Q&A