Cole-Parmer Cyclone & Collection Bin for Industrial Fluidized Sand Baths
Representative image only.
Cole-Parmer Cyclone & Collection Bin for Industrial Fluidized Sand Baths
Cole-Parmer | Item: 0118480
₹655,000.00INR / Each
₹655,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Cyclone & Collection Bin for Industrial Fluidized Sand Baths
Cole-Parmer Cyclone & Collection Bin for Industrial Fluidized Sand Baths
0118480
₹655,000.00INR / Each
+ + + =
Total Price 1334000.00 ADD 4 ITEMS TO CART
Product Description
Key Features
  • Collect aluminum oxide for reuse
  • 3 year warranty
  • Previous part number FSCN1
More About this Item
Parts and accessories to meet your application needs and keep your sand bath operating dependably.
Wall-mount unit extracts all the aluminum oxide that are carried into the bath exhaust system when the extraction fan is in use.
3 Year Warranty
Specifications
  • Compatible WithIndustrial fluidized sand baths 01184-10 and 01184-20
  • DescriptionCyclone & Collection Bin for Industrial Fluidized Sand Baths