Cole-Parmer Cuvette Holder, 10 x 10 mm Square
Representative image only.
Cole-Parmer Cuvette Holder, 10 x 10 mm Square
Cole-Parmer | Item: 8307041
Optimize performance of your Cole-Parmer® spectrophotometer
Call For Price
Call For Price
Product Description
Key Features
  • Expand analysis capabilities or make testing easier
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
The single-position cell/cuvette holder is compatible with the 63, 72, 73, 74, and 76 series, as well as the Genova Bio and Plus spectrophotometers. This holder accepts 10 x 10 mm cuvettes for a 10 mm pathlength.
Specifications
  • Compatible WithCole-Parmer SP-200,SP-350, SP-400, SP-500, SP-600 Spectrophotometers
  • DescriptionCuvette Holder, 10 x 10mm Square