Cole-Parmer Cryovial Rack, 40, Red; 1/Ea
Representative image only.
Cole-Parmer Cryovial Rack, 40, Red; 1/Ea
Cole-Parmer | Item: 0439572
Effortlessly open vials with one hand
₹5,400.00INR / Each
₹5,400.00INR / Each
Product Description
Key Features
  • Constructed of autoclavable polypropylene
  • Suitable for a temperature range of -196°C to +121°C
More About this Item
Securely holds up to 40 self-standing cryogenic vials. Unique locking design of wells allows for one-handed opening and closing of caps.
Specifications
  • Tubes Held40
  • Lowest Set Temperature (° C)-196
  • ColorRed
  • DescriptionPolarSafe Cryovial Rack, 40, Red; 1/Ea