Cole-Parmer Cryovial Cap Insert, White; 100/Pk
Representative image only.
Cole-Parmer Cryovial Cap Insert, White; 100/Pk
Cole-Parmer | Item: 0439575
Easily identify your samples with color-coded cap inserts
₹1,300.00INR / Pkg of 100
₹1,300.00INR / Pkg of 100
Product Description
Key Features
  • Choose from 6 color options or an assortment pack!
  • New design with larger cap inserts for more surface area
More About this Item
Cap inserts are available in assorted colors and snap easily into place on both internally and externally threaded caps. Ideal for color coding and sample identification.
Specifications
  • ColorWhite
  • DescriptionCryovial Cap Insert, White; 100/Pk