Cole-Parmer Cryocane Coders, Metal, Blue; 12/Pk
Representative image only.
Cole-Parmer Cryocane Coders, Metal, Blue; 12/Pk
Cole-Parmer | Item: 2441009
Easily identify cryogenic storage canes
₹3,500.00INR / Pkg of 12
₹3,500.00INR / Pkg of 12
Product Description
Key Features
  • Ideal for samle identifical of Cole-Parmer CryoCanes
  • Costructed of durable metal
  • Easily clip onto the end of your cryocane for a secure fit
More About this Item
Unique, colorful tabs keep track of your inventory of CryoCanes. Identify a particular batch of vials or a specific technician's work. Coder tabs remain firmly in place during storage in liquid-nitrogen freezers.
Specifications
  • ColorBlue
  • DescriptionCryocane Coders, Metal, Blue; 12/Pk