Cole-Parmer Cryo-Sentry Low-Level Alarm for Dewars 03773-08 and -09
Representative image only.
Cole-Parmer Cryo-Sentry Low-Level Alarm for Dewars 03773-08 and -09
Cole-Parmer | Item: 0377318
A visual and an audible alarms alert you of low liquid nitrogen levels
₹410,000.00INR / Each
₹410,000.00INR / Each
Product Description
Key Features
  • Automatically checks the liquid level every 30 minutes
  • Connect an external alarm device to the relay output
  • Operates on AA batteries or AC power
  • Use with dewar 03773-08 and -09
  • 3 month warranty
More About this Item
Protect your valuable biological samples with the low-level alarm for your cryogenic storage dewars. The low-level alarm checks the liquid level every 30 minutes conserving battery power. Easily check the level without opening the vessel by pressing the LEVEL CHECK button.
3 Month Warranty
Specifications
  • Compatible WithDewars 03773-08 and -09
  • Battery4 AA
  • Power (VDC)120
  • DescriptionCryo-Sentry Low-Level Alarm for Dewars 03773-08 and -09
  • Warranty3 months