Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 quart
Representative image only.
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 quart
Cole-Parmer | Item: 7975500
Ensures optimum pump performance
₹2,600.00INR / Quart of 1
₹2,600.00INR / Quart of 1
Buy It Together
Current Item
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 quart
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 quart
7975500
₹2,600.00INR / Quart of 1
+
Edwards Vibration isolator (pack of 4) for E2M28
7930061
₹57,300.00INR / Pkg of 4
=
Total Price 59900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general purposes
More About this Item
Vacuum pump oil is formulated to ensure optimum pump performance and pump life.
CP 500 is a general-purpose high-viscosity oil. An excellent custom blended, general purpose hydrocarbon fluid utilizing advanced feedstocks. This rotary vane pump fluid is a lower cost alternative to molecularly distilled fluids. It is useful where frequent fluid changes cause large volumes of oils to be used.
Discounts: Save 10% on 12-23 quarts, 15% on 24-35 quarts and 20% on 36 or more quarts. Save 10% on 4-11 gallons, 15% on 12-23 gallons and 20% on 24 or more gallons.
Specifications
  • FormLiquid
  • Viscosity (cSt)68
  • Volume (mL)946
  • Volume (Qt)1
  • Volume (Gal)0.25
  • Vapor Pressure (torr)0.0002 at 25°C
  • Packaging946 mL
  • DescriptionVacuum Pump Oil, 68 cSt, 1 quart