Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 gal
Representative image only.
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 gal
Cole-Parmer | Item: 7975502
Ensures optimum pump performance
₹9,700.00INR / Gallon of 1
₹9,700.00INR / Gallon of 1
Buy It Together
Current Item
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 gal
Cole-Parmer CP 500 Vacuum Pump Oil, 68 cSt, 1 gal
7975502
₹9,700.00INR / Gallon of 1
+
Edwards Vibration isolator (pack of 4) for E2M28
7930061
₹57,300.00INR / Pkg of 4
=
Total Price 67000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ideal for general purposes
More About this Item
Vacuum pump oil is formulated to ensure optimum pump performance and pump life.
An excellent custom blended, general purpose hydrocarbon fluid utilizing advanced feedstocks. This rotary vane pump fluid is a lower cost alternative to molecularly distilled fluids. It is useful where frequent fluid changes cause large volumes of oils to be used.
Discounts: Save 10% on 12-23 quarts, 15% on 24-35 quarts and 20% on 36 or more quarts. Save 10% on 4-11 gallons, 15% on 12-23 gallons and 20% on 24 or more gallons.
Specifications
  • FormLiquid
  • Viscosity (cSt)68
  • Volume (mL)3785
  • Volume (Qt)4
  • Volume (Gal)1
  • Vapor Pressure (torr)0.0002 at 25°C
  • Packaging3785 mL
  • DescriptionVacuum Pump Oil, 68 cSt, 1 gal