Cole-Parmer CP 19 Vacuum Pump Oil, 55 cSt, 1 gallon
Representative image only.
Cole-Parmer CP 19 Vacuum Pump Oil, 55 cSt, 1 gallon
Cole-Parmer | Item: 7975512
Ensures optimum pump performance
₹9,700.00INR / Gallon of 1
₹9,700.00INR / Gallon of 1
Buy It Together
Current Item
Cole-Parmer CP 19 Vacuum Pump Oil, 55 cSt, 1 gallon
Cole-Parmer CP 19 Vacuum Pump Oil, 55 cSt, 1 gallon
7975512
₹9,700.00INR / Gallon of 1
+
Edwards Vibration isolator (pack of 4) for E2M28
7930061
₹57,300.00INR / Pkg of 4
=
Total Price 67000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Resist gum formation and oxidation
More About this Item
Vacuum pump oil is formulated to ensure optimum pump performance and pump life.
Use CP 19 in direct drive pumps to resist gum formation and oxidation. The standard of general purpose hydrocarbon oils, the CP-19 is recommended for use in non-corrosive applications. Through molecular distillation, light end fractions are removed, resulting in lower vapor pressures and reduced backstreaming compared to undistilled refinery products. These properties allow CP-19 to be used with the more expensive diffusion pump fluids that can otherwise be contaminated by a lesser fluid. Compared to refinery fluids, CP-19 offers reduced maintenance costs, better pumping cycles, longer fluid life, and a cooler running pump.
Discounts: Save 10% on 12-23 quarts, 15% on 24-35 quarts and 20% on 36 or more quarts. Save 10% on 4-11 gallons, 15% on 12-23 gallons and 20% on 24 or more gallons.
Specifications
  • FormLiquid
  • Viscosity (cSt)55
  • Volume (mL)3785
  • Volume (Qt)4
  • Volume (Gal)1
  • Vapor Pressure (torr)3 x 10 -5 25°C
  • Packaging3785 mL
  • DescriptionVacuum Pump Oil, 55 cSt, 1 gallon