Cole-Parmer Cork Borer, 12 Piece Set, Plated Brass
Representative image only.
Cole-Parmer Cork Borer, 12 Piece Set, Plated Brass
Cole-Parmer | Item: 0629899
Easily convert solid stopper into holed stopper
₹13,500.00INR / Each
₹13,500.00INR / Each
Product Description
Key Features
  • Steel punch makes boring easier
  • 1 year warranty
More About this Item
Convert your solid stopper into holed-stopper with these easy-to-use borer sets. All borers come with a steel punch to make boring easier. Borer sharpener (sold separately as part number 06298-94) keeps your borer in top condition and sharpens borers up to 25mm in diameter.
Boring diameter 4 to 22 mm
1 Year Warranty
Specifications
  • MaterialBrass
  • DescriptionCork Borer, 12 Piece Set, Plated Brass