Cole-Parmer Contact Tachometer Linear Speed Wheel, 12" Dia
Representative image only.
Cole-Parmer Contact Tachometer Linear Speed Wheel, 12" Dia
Cole-Parmer | Item: 0820361
Enhance performance of your Cole-Parmer® Contact Tachometer
₹15,100.00INR / Each
₹15,100.00INR / Each
Buy It Together
Current Item
Cole-Parmer Contact Tachometer Linear Speed Wheel, 12" Dia
Cole-Parmer Contact Tachometer Linear Speed Wheel, 12" Dia
0820361
₹15,100.00INR / Each
+
Cole-Parmer Contact Tachometer with LCD Display
0820450
₹81,100.00INR / Each
=
Total Price 96200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Measure the speed of motors, conveyor belts, and other moving and rotational systems
  • 1 year warranty
More About this Item
Compatible for use with tachometer models 08204-50, 08204-80, 23022-57, 54500-97, 87304-10, and 98807-37; tachometer sold separately.
1 Year Warranty
Specifications
  • Compatible WithTachometers 08204-50, 08204-80, 23022-57, 54500-97, 87304-10, and 98807-37
  • DescriptionContact Tachometer Linear Speed Wheel, 12" Dia