Cole-Parmer Conductivity Probe, 2-cell, K = 10.0, Epoxy Body/Pt Bands, 2.5-ft Cable
Representative image only.
Cole-Parmer Conductivity Probe, 2-cell, K = 10.0, Epoxy Body/Pt Bands, 2.5-ft Cable
Cole-Parmer | Item: 3560878
A variety of conductivity probes for any application
₹26,000.00INR / Each
₹26,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Conductivity Probe, 2-cell, K = 10.0, Epoxy Body/Pt Bands, 2.5-ft Cable
Cole-Parmer Conductivity Probe, 2-cell, K = 10.0, Epoxy Body/Pt Bands, 2.5-ft Cable
3560878
₹26,000.00INR / Each
+ + + + + + + +
Traceable Calibrated Portable Conductivity Meter
1960103
₹72,600.00INR / Each
=
Total Price 737100.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose a cell constant compatible with your application
  • Varying cable lengths available
  • Replacement Conductivity Cell for Oakton models CON 700, CON 2700 series meters
  • 1 year warranty
1 Year Warranty
Specifications
  • Cell Constant10
  • Body MaterialEpoxy
  • Sensor MaterialPlatinum
  • Use WithEnvironmental Express Oakton® CON 700 and CON 2700 meters
  • Cable Length (ft)2.5
  • Cell Configuration2 cell
  • DescriptionConductivity Probe, 2-cell, K = 10.0, Epoxy Body/Pt Bands, 2.5-ft Cable