Cole-Parmer Comfort-Grip Motorized Pipette Controller, 110/220 VAC
Representative image only.
Cole-Parmer Comfort-Grip Motorized Pipette Controller, 110/220 VAC
Cole-Parmer | Item: 2520015
Comfortable grip and wireless operation combined in an affordable package
₹116,000.00INR / Each
₹116,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer Comfort-Grip Motorized Pipette Controller, 110/220 VAC
Cole-Parmer Comfort-Grip Motorized Pipette Controller, 110/220 VAC
2520015
₹116,000.00INR / Each
+ =
Total Price 123700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
0.2 µm filter, rechargeable battery, and universal voltage battery charger.
Key Features
  • Dial in the perfect pipetting speed with the adjustable thumbwheel
  • In-line membrane filter prevents aerosol and liquid contamination
  • Silicone nosepiece is removable and autoclavable
  • Rechargeable battery provides eight hours of continuous operation
  • Check charge status via red and green LED indicators
  • 1 year warranty
More About this Item
This pipette controller will work with any standard pipette up to 100 mL—plus the ergonomic design is ideal for extended use. Pump speed is adjustable continuously rather than in discrete increments, and can be changed during operation. There are two LED charge status indicators, green and red. Green indicates the battery is charged and red means the battery is low.
1 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Dispensing (mL)100
  • Overall Length (InchShort)8.75
  • Overall Length (mm)222.25
  • Filter Type1x 0.2µm (option of either 0.2µm or 0.45µm filters available)
  • DescriptionMotorized Pipette Controller, 110/220 VAC