Cole-Parmer Column Clamp, Small, Stainless Steel, 2.56" to 3.5" Grip Range
Representative image only.
Cole-Parmer Column Clamp, Small, Stainless Steel, 2.56" to 3.5" Grip Range
Cole-Parmer | Item: 0806010
Provide support for your specific laboratory equipment
₹23,900.00INR / Each
₹23,900.00INR / Each
Product Description
Key Features
  • Durable stainless steel clamps offer chemical resistance and increased strength
  • 1 year warranty
More About this Item
Ideal for large cylindrical glassware and similar labware. The large, flat thumbscrew/worm-drive provides a firm hold. NOTE: An additional connector must be purchased in order to attach clamps to frames or other apparatus.
1 Year Warranty
Specifications
  • Clamp MaterialStainless Steel
  • Clamp TypeEquipment, Column
  • Min Grip Range (InchShort)2.56
  • Max Grip Range (InchShort)3.5
  • Length (in)11.1
  • DescriptionColumn Clamp, Small, Stainless Steel, 2.56" to 3.5" Grip Range