Cole-Parmer Cleanroom Mop with Propylene Handle

Representative image only.

Cole-Parmer Cleanroom Mop with Propylene Handle

Cole-Parmer | Item: 1597248

Designed for controlled environments from Class 100 to Class 100,000 cleanrooms

₹30,500.00INR / Box of 1

₹30,500.00INR / Box of 1
Product Description

Includes

Each box contains - 2 Flat Mop frame+ 2 Telescopic handle+10pcs of mop cover

Key Features

  • High-density microfiber mop cover with ultra dust absorbency Mop head has a unique “butterfly clip’’ design which improves cleaning performance greatly and prevents scratching Universal rotating design lets you access hard-to-reach areas of ceilings, walls, and other confined areas under equipment Interlocking handle structure allows more flexibility, stability, and adjustments to the mop length when necessary Contains no hazardous substances such as lead, cadmium, mercury, chromium VI, etc
Specifications
  • MaterialPropylene Handle
  • Length (in)24 51/64
  • Depth (in)8.3
  • Width (cm)21
  • Length (cm)63
  • DescriptionCleanroom Mop with Propylene handle; 1set/Bx
Reviews
Q&A