Cole-Parmer Chromatography Column, 40 mL nominal volume

Representative image only.

Cole-Parmer Chromatography Column, 40 mL nominal volume

Cole-Parmer | Item: 3462410

₹182,000.00INR / Case of 1

₹182,000.00INR / Case of 1
Product Description

Key Features

  • Chromatography columns with PTFE valve, coarse fritted disc, and flow control adapter

More About this Item

Low pressure (7 psig max) medium wall chromatography column with reservoir. Columns feature a PTFE insert with a locknut that attaches the flow control valve to the column. Top closure is a GL-45 thread for a positive connection. An all PTFE-valve and exposed drip lip eliminate tip damage. Columns include bottom valve assemblies, GL-45 caps, PTFE inserts, lock-nuts, O-rings, and flow adapters.

Specifications
  • MaterialGlass
  • Capacity (mL)40
  • DescriptionChromatography Column, 40 mL nominal volume
Reviews
Q&A