Cole-Parmer Chloride Standard for Chloride Analyzer 02656-20; 1 x 100 mL Bottle
Representative image only.
Cole-Parmer Chloride Standard for Chloride Analyzer 02656-20; 1 x 100 mL Bottle
Cole-Parmer | Item: 0265639
Use the standard solution to ensure accurate and reliable measurements
₹11,200.00INR / Each
₹11,200.00INR / Each
Product Description
Key Features
  • 200 mg/L concentration
  • 100 mL bottle
Specifications
  • Compatible WithChloride analyzer 02656-20
  • DescriptionChloride Standard, 1 x 100 mL bottle