Cole-Parmer Cell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs
Representative image only.
Cole-Parmer Cell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs
Cole-Parmer | Item: 0439601
Secure single-cell suspensions and remove debris
₹16,400.00INR / Case of 50
₹16,400.00INR / Case of 50
Buy It Together
Current Item
Cole-Parmer Cell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs
Cole-Parmer Cell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs
0439601
₹16,400.00INR / Case of 50
+ =
Total Price 31700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Pair directly with your 50-mL centrifuge tubes
  • Color-coded frames help differentiate sizes
  • Extended tab makes handling easy
  • Individually packaged and gamma sterilized
  • DNase-& RNase-free
  • Non-pyrogenic
  • 70 µm mesh pore size
More About this Item
These Cole-Parmer® Essentials cell strainers are sterile, DNase- and RNase free, and non-pyrogenic, ideal for protecting your valuable flow cytometry and cell sorting instrumentation. Cell strainers are easy-to-use devices for isolating primary cells to consistently obtain a uniform single-cell suspension from tissues. Reliably remove clumps and debris from cell suspensions and clinical samples prior to analysis. Simply put cell strainer directly on 50 mL centrifuge tube for easy, immediate filtration.
Cell strainers are constructed from a strong nylon mesh with evenly spaced mesh pores and a durable polypropylene frame. Color-coded frames made for quick identification of different pore sizes. The extended lip on the strainer enables aseptic handling with forceps.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
Bio DNase-FreeNonpyrogenicRNase-FreeSterile
Specifications
  • Membrane MaterialNylon
  • Pore Size (micron)70
  • Housing MaterialPolypropylene
  • SterileYes
  • DescriptionCell Strainer, Sterile, 70 µm, White, Individually Wrapped; 50/Cs