Cole-Parmer Cell Lifters, Sterile, HDPE, Flat and Narrow Blades; 100/Cs
Representative image only.
Cole-Parmer Cell Lifters, Sterile, HDPE, Flat and Narrow Blades; 100/Cs
Cole-Parmer | Item: 0195914
Double-ended cell lifter design provides two tools in one for quick removal of cell layers
₹44,900.00INR / Case of 100
₹44,900.00INR / Case of 100
Product Description
Key Features
  • Easily harvest cells from culture dishes, plates, and tubes
  • 19 mm beveled edge flat blade ensures even contact with growth surface
  • Sterilized by gamma irradiation
  • Individually packaged
  • Tapered narrow blade fits into 96-well plates or microcentrifuge tubes
More About this Item
Double-ended lifters allow easy harvest of cells from culture dishes, plates and tubes. Both versions have a 19 mm beveled edge flat blade on one end. Second end choices include a narrow-tapered blade for 96-well plates or microcentrifuge tubes, or a J-hook to vertically lift cells from 4- to 48-well plates. Made of high-density polyethylene, these cell lifters are individually wrapped and sterilized by gamma irradiation, and are RNase/DNase free, nonpyrogenic, and noncytotoxic.
Features a 19 mm beveled edge flat blade to ensure even contact with growth surface of flasks and dishes on one end, and a narrow blade tapered to fit into 96-well plates or microcentrifuge tubes on the other. The narrow blade can also be used to resuspend stubborn pellets.
Sterile
Specifications
  • SterileYes
  • Overall Length (mm)230
  • Overall Length (in)9
  • ColorWhite
  • MaterialHDPE
  • DescriptionCell Lifters, Sterile, HDPE, Flat and Narrow Blades; 100/Cs