Cole-Parmer Carbon Dioxide Meter/Controller Calibration Kit
Representative image only.
Cole-Parmer Carbon Dioxide Meter/Controller Calibration Kit
Cole-Parmer | Item: 2900070
Optimize performance of your CO2 meter
₹13,800.00INR / Each
₹13,800.00INR / Each
Product Description
Key Features
  • Compatible for use with your 56105-00 CO2 meter
  • Max pressure 45 psi (3.1 bar)
  • PEEK body and ferrule/PK nut construction
  • 0.006" (0.150 mm) thru-hole diameter
  • 1 year warranty
More About this Item
This calibration kit includes two standards, Standard A for zero point calibration and Standard B is the reference or slope calibration.
ISA and must be ordered seperatly.
1 Year Warranty
Specifications
  • Compatible With56105-00
  • DescriptionCarbon Dioxide Meter/Controller Calibration Kit