Cole-Parmer Bottletop Dispenser, Recirculation Valve, 2.5 to 30 ML

Representative image only.

Cole-Parmer Bottletop Dispenser, Recirculation Valve, 2.5 to 30 ML

Cole-Parmer | Item: 2520834

Save money by conserving reagents

₹86,300.00INR / Each

₹86,300.00INR / Each
Product Description

Includes

discharge tube, telescoping filling tube, calibration tool, bottle adapter set (28, 32, 38, 40, and 45 mm), and calibration certificate.

Key Features

  • Recirculation valve prevents loss of reagent during priming and purging
  • Smooth plunger movement provides bubble-free dispensing
  • Adjustable nozzle allows you to change the dispensing angle
  • 1 year warranty
  • New, improved version of PD30R

More About this Item

These bottletop dispensers conserve reagents with a built-in recirculation valve allowing you to prime and purge without waste. The PTFE piston with ETP o-ring provide frictionless movement reducing the occurrence of bubbles in your dispenses. The springless PTFE valve manifold offers superior chemical compatibility and resists jamming.

1 Year WarrantyConformite Europeenne
Specifications
  • Min Dispense Volume (mL)2.5
  • Max Dispense Volume (mL)30
  • Division (mL)0.5
  • Accuracy±0.6%
  • Reproducibility±0.2%
  • AutoclavableYes
  • Wetted PartsPTFE, ETP, borosilicate glass, PP, PFA, FEP
  • DescriptionBottletop Dispenser, 2.5 to 30 mL
  • Warranty1 year
Reviews
Q&A