Cole-Parmer Beaker Positioning Cover for 1.5 Gal Ultrasonic Cleaners, 2 x 600 mL Beakers
Representative image only.
Cole-Parmer Beaker Positioning Cover for 1.5 Gal Ultrasonic Cleaners, 2 x 600 mL Beakers
Cole-Parmer | Item: 0889826
Achieve optimal cleaning results
₹17,700.00INR / Each
₹17,700.00INR / Each
Product Description
Includes
Beaker cover and retaining o-rings.
Key Features
  • Suspends beakers for increased ultrasonic activity
  • Ideal for cleaning small parts
  • Prevents damage to transducers
  • Holds 2 x 600 mL Beakers
More About this Item
Use these beaker positioning covers with beakers to clean small objects or for indirect cleaning. Cover suspends beakers above tank bottom for increased ultrasonic activity.
Ultrasonic activity is greatest about an inch from the bottom of the tank, so suspending your items provides the best cleaning possible and helps prevent damage to the transducers.
Order beakers separately.
Please Note: Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners.
Specifications
  • Capacity (Gal)1.5
  • Capacity (Liters)5.68
  • MaterialPolypropylene
  • DescriptionBeaker Positioning Cover for 1.5 Gal Ultrasonic Cleaners, 2 x 600 mL Beakers