Cole-Parmer Beaker and Vessel Holder Kit
Representative image only.
Cole-Parmer Beaker and Vessel Holder Kit
Cole-Parmer | Item: 2500001
Bundled kit for convenience and savings
₹80,900.00INR / Pkg of 1
₹80,900.00INR / Pkg of 1
Product Description
Includes
lab stand, support rod, chain clamp, and clamp holder.
Key Features
  • Mount your overhead mixer directly to the 18" long, 1/2" diameter support rod
  • Place your vessel on the 6" x 9" support base, and use the chain clamp to secure it in place
  • Works to support vessels on magnetic stirrers, too!
More About this Item
We've paired our most popular lab stand, support rod, and vessel clamp for your benefit. The stainless steel lab support rod and cast-iron support base provide maximum strength and durability so you can mount your overhead laboratory mixer with confidence. The chain clamp has a quick slip-on-chain connection and a large easy-to-turn adjustment knob. The flexible chain is able to hold circular and irregularly shaped apparatus, up to an 11" (28 cm) diameter, firmly in place while mixing.
Specifications
  • MaterialStainless Steel rod, cast-iron base, nickel-plated holder and chain-clamp
  • DescriptionBeaker and Vessel Holder Kit