Cole-Parmer Autoclave Deodorizers, Apple; 40/Bottle
Representative image only.
Cole-Parmer Autoclave Deodorizers, Apple; 40/Bottle
Cole-Parmer | Item: 2440675
Absorb and remove autoclave odors rather than just covering them
₹7,600.00INR / Pkg of 40
₹7,600.00INR / Pkg of 40
Product Description
Key Features
  • Choose apple or lemon scented capsules
More About this Item
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture dissolves the capsule and releases a clean, fresh scent.
Specifications
  • DescriptionAutoclave Deodorizers, Apple; 40/Bottle