Cole-Parmer ATS10232E Multi-Input Light Measurement Module for Integrity 10
Representative image only.
Cole-Parmer ATS10232E Multi-Input Light Measurement Module for Integrity 10
Cole-Parmer | Item: 3663089
Keep your reaction station running smoothly with these parts and accessories
₹478,000.00INR / Each
₹478,000.00INR / Each
Product Description
Key Features
  • Suitable for use with Cole-Parmer® Integrity 10, RS-200, or RS-250 series reaction stations
  • Accommodates 10 infrared probes
  • 1 year warranty
  • Previous part number ATS10232E
More About this Item
Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
The module features 10 multicoloured LED indicators (one per channel). Flashing red indicates channel off, green means channel clear, and orange means channel turbid. Opitcal wavelength is 940nm.
1 Year Warranty
Specifications
  • Compatible WithCole-Parmer Integrity 10 Reaction Station
  • DescriptionMulti-Input Light Measurement Module for Integrity 10