Cole-Parmer ATS10001 Multi-Temp 10 Plug-In Box for Integrity 10
Representative image only.
Cole-Parmer ATS10001 Multi-Temp 10 Plug-In Box for Integrity 10
Cole-Parmer | Item: 3663080
Keep your reaction station running smoothly with these parts and accessories
₹551,000.00INR / Each
₹551,000.00INR / Each
Product Description
Key Features
  • Suitable for use with Cole-Parmer® Integrity 10, RS-200, or RS-250 series reaction stations
  • Accommodates 10 thermocouple probes
  • 1 year warranty
  • Previous part number ATS10001
More About this Item
Choose from a wide range of accessories suitable for your reaction station including reaction vessels, probes, and software.
Provides multiple temperature sensor input capability, allowing up to 12 thermocouple sensors to be monitored simultaneously. Output is via standard RS232 interface.
1 Year WarrantyConformite Europeenne
Specifications
  • Compatible WithCole-Parmer Integrity 10 Reaction Station
  • DescriptionMulti-Temp 10 Plug-In Box for Integrity 10