Cole-Parmer Aseptic Sampling Cap for GL45 Bottles, 2 Ports, 1/8"ID x 1/4"OD TPE Tubing; 1/Ea
Representative image only.
Cole-Parmer Aseptic Sampling Cap for GL45 Bottles, 2 Ports, 1/8"ID x 1/4"OD TPE Tubing; 1/Ea
Cole-Parmer | Item: 5014250
Specifically designed for your existing glass media bottles
₹16,000.00INR / Each
₹16,000.00INR / Each
Product Description
Key Features
  • Closed system protects sample integrity
  • Eliminate cleaning validation
  • Reduce assembly and installation time
  • Two ports with 1/8" ID x 1/4" OD TPE tubing vent and transfer lines
  • USP Class VI and RoHS compliant
More About this Item
The one-piece molded closure system is made with polypropylene cap and TPE tubing. Designed to work with GL45 glass media bottles, the cap provides a closed system to reduce contamination and help protect sample integrity. Cap features two ports: one for venting and one for transferring. Venting port comes with a 4"L tube; transferring port comes with an 18"L tube inside the bottle and an 18"L tube outside the bottle. Install on your glass bottle and autoclave before use.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Cap MaterialPolypropylene
  • Cap / Stopper SizeGL45
  • Cap ColorWhite
  • Special FeaturesTwo ports: venting and transferring with 1/8"ID x 1/4"OD TPE tubing
  • AutoclavableYes
  • DescriptionAseptic Sampling Cap for GL45 Bottles, 2 Ports, 1/8"ID x 1/4"OD TPE Tububing; 10/Pk