Cole-Parmer Aluminum Freezer Rack Tags, with Number; Yellow
Representative image only.
Cole-Parmer Aluminum Freezer Rack Tags, with Number; Yellow
Cole-Parmer | Item: 2441032
Customize or upgrade your freezer rack
₹4,100.00INR / Each
₹4,100.00INR / Each
Product Description
Key Features
  • Constructed of durable aluminum
  • Easily identify with yellow color
More About this Item
Suitable for use with your PolarSafe freezer racks.
These aluminum tags are ideal for use with upright freezer, chest freezer, and liquid nitrogen tank racks. Tags are 2” in diameter and include a 1” number engraved in black. Please include number selection when ordering.
Specifications
  • MaterialAluminum
  • ColorYellow
  • Compatible WithUpright freezer, chest freezer, and liquid nitrogen tank racks
  • DescriptionAluminum Freezer Rack Tags, with Number; Yellow