Cole-Parmer All-in-One pH/ATC Electrode for 600 series meters, Double-Junction
Representative image only.
Cole-Parmer All-in-One pH/ATC Electrode for 600 series meters, Double-Junction
Cole-Parmer | Item: 3581672
Get accurate readings for even demanding applications
₹25,300.00INR / Each
₹25,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer All-in-One pH/ATC Electrode for 600 series meters, Double-Junction
Cole-Parmer All-in-One pH/ATC Electrode for 600 series meters, Double-Junction
3581672
₹25,300.00INR / Each
+ =
Total Price 45600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Choose a pH electrode based on your application
  • For clean samples, measure pH with single-junction electrodes
  • Use double-junction electrodes for samples with contaminations or particulates, heavy metals, or organic compounds.
  • For pH 0 to 12 readings
  • Compatible with dirty samples or solutions containing heavy metals or organics
  • 1 year warranty
More About this Item
Low-maintenance epoxy-bodied electrodes provide the durability for field
work. Refillable electrodes provide greater accuracy and faster response for
more critical analytical work.
1 Year Warranty
Specifications
  • MaterialEpoxy
  • Compatible WithOakton 600 series meters
  • DescriptionAll-in-One pH/ATC Electrode for 600 series meters, Double-Junction