Cole-Parmer Adaptor for Express 445 Gas Cartridges
Representative image only.
Cole-Parmer Adaptor for Express 445 Gas Cartridges
Cole-Parmer | Item: 0439720
Enhance the performance of your Cole-Parmer® burners
₹27,600.00INR / Each
₹27,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer Adaptor for Express 445 Gas Cartridges
Cole-Parmer Adaptor for Express 445 Gas Cartridges
0439720
₹27,600.00INR / Each
+
Cole-Parmer Bunsen Burner XT with IR Sensor
0439717
₹192,000.00INR / Each
=
Total Price 219600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Quickly connect gas cartridges to your Bunsen Burner
More About this Item
A variety of fuel adapters and holders to accommodate different burners and applications.
Compatible with Bunsen burners models 04397-17 and -18. Burners sold separately. This adapter is suitable for use with the Express 445 Gas Cartridges, which Cole-Parmer does not offer due to hazardous restrictions at this time.
04397, 04397-1, 04397-2, 04397 1, 04397 2, EW, GH, SI, AO
Specifications
  • DescriptionGas Cartridge Adaptor for Express 445