Cole-Parmer Adapter, single pin DIN meter to BNC electrode
Representative image only.
Cole-Parmer Adapter, single pin DIN meter to BNC electrode
Cole-Parmer | Item: 0598321
₹14,900.00INR / Each
₹14,900.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
BNC is the most common electrode connector, but some pH meters and electrodes come with different types of connectors. Order an adapter to ensure your meter and electrode are compatible.
1 Year Warranty
Specifications
  • DescriptionAdapter, single pin DIN meter to BNC electrode