Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Shallow Well, Round, Pre-Slit; 5/PK
Representative image only.
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Shallow Well, Round, Pre-Slit; 5/PK
Cole-Parmer | Item: 1292019
Ensure purity and prevent loss
₹9,300.00INR / Pkg of 5
₹9,300.00INR / Pkg of 5
Product Description
Key Features
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 96-well, shallow round, pre-slit
More About this Item
Use these microplate sealing mats with well plates for storage prior to analysis. Silicone reseals after injections and pipetting for contamination-free sample preservation. Mats fit standard 96- and 384-well microplates.
Specifications
  • Number of Wells96
  • Well ShapeRound, shallow
  • MaterialSilicone
  • Description96-Well Microplate Sealing Mat, Silicone, Shallow Well, Round, Pre-Slit; 5/PK