Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Round, Low Bleed; 5/PK
Representative image only.
Cole-Parmer 96-Well Microplate Sealing Mat, Silicone, Round, Low Bleed; 5/PK
Cole-Parmer | Item: 1292017
Ensure purity and prevent loss
₹10,500.00INR / Pkg of 5
₹10,500.00INR / Pkg of 5
Product Description
Key Features
  • Silicone reseals effectively to maintain sample purity
  • Individual well plugs eliminate cross-contamination
  • Pre-slit option available for easy pipetting
  • 96-well, low bleed, round, no slit
More About this Item
Use these microplate sealing mats with well plates for storage prior to analysis. Silicone reseals after injections and pipetting for contamination-free sample preservation. Mats fit standard 96- and 384-well microplates.
Specifications
  • Number of Wells96
  • Well ShapeRound, deep
  • MaterialLow-bleed silicone
  • Description96-Well Microplate Sealing Mat, Silicone, Round, Low Bleed; 5/PK