Cole-Parmer 5/100-09-BX Replacement Filter Tubes 5/Pk
Representative image only.
Cole-Parmer 5/100-09-BX Replacement Filter Tubes 5/Pk
Cole-Parmer | Item: 0291753
₹61,300.00INR / Box of 5
₹61,300.00INR / Box of 5
Product Description
More About this Item
Replacement tubes for 02917-XX series filters. Remove oil, water, and solid contaminants.
Specifications
  • Compatible WithParker Hannifin 8B02N-OA2-BX Compressed-Air Filter (02917-10)
  • DescriptionReplacement Filter Tubes 5/bx