Cole-Parmer 3.SA.DN.6 Foam Grip Tweezers w/ Long, Very Sharp, Fine Tips
Representative image only.
Cole-Parmer 3.SA.DN.6 Foam Grip Tweezers w/ Long, Very Sharp, Fine Tips
Cole-Parmer | Item: 0738745
Electrically static dissipative, ESD-safe material
₹11,300.00INR / Each
₹11,300.00INR / Each
Product Description
Key Features
  • Soft PVC foam-grip handle provides relief from hand fatigue
  • Antimagnetic and anti-acid stainless steel tips
  • Excellent chemical and wear resistance
  • Limited Lifetime Warranty
More About this Item
Choose the tip style that works for your application.
Limited Lifetime Warranty
Specifications
  • Overall Length (in)5
  • Overall Length (cm)12.5
  • MaterialStainless Steel w/ ESD Foam Handles
  • SterileNo
  • DescriptionFoam Grip Tweezers w/ Long, Very Sharp, Fine Tips
  • WarrantyLimited Lifetime