Cole-Parmer 1 to 1500 torr Pressure/Vacuum Diaphragm-Type Sensor, 1/8" NPT(M)
Representative image only.
Cole-Parmer 1 to 1500 torr Pressure/Vacuum Diaphragm-Type Sensor, 1/8" NPT(M)
Cole-Parmer | Item: 6880157
Create your own high-accuracy system to monitor or control your application
₹108,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹108,000.00INR / Each
Buy It Together
Current Item
Cole-Parmer 1 to 1500 torr Pressure/Vacuum Diaphragm-Type Sensor, 1/8" NPT(M)
Cole-Parmer 1 to 1500 torr Pressure/Vacuum Diaphragm-Type Sensor, 1/8" NPT(M)
6880157
₹108,000.00INR / Each
+ + =
Total Price 273700.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Required system components include the sensor and indicator or controller all sold separately
  • 2 year warranty
More About this Item
Assemble a complete pressure/vacuum system that meets your monitoring needs. First choose a sensor with the range appropriate for your application. Then order an indicator or a controller—both are available in six ranges. Complete your system with a compatible calibrator to fine tune your meter or controller.
Sensors provide improved tracking ability and maintain calibration over long periods of time. When replacing a sensor, you do not need to recalibrate. These sensors are suited for use with mercury and oil pumps.
Indicators provide precise digital readout of pressure/vacuum.
Controllers have all of the features of the indicators plus two independently set SPDT relays for control of high and low limits. Easily adjust set points to any value with front-panel push buttons; connect relay outputs at screw terminals at back of unit.
Calibrators let you readjust your meter or controller display quickly and easily to factory specifications
Two year warranty
Specifications
  • Accuracy±1% of reading or ±1 Torr
  • Wetted MaterialsSilicon
  • Process Connection1/8" NPT(M)
  • Media CompatibilityGases compatible with wetted materials
  • Resolution1 Torr
  • Description1 to 1500 torr Pressure/Vacuum Diaphragm-Type Sensor, 1/8" NPT(M)