Clavies Biohazard Bag Stand with Tray for 10 to 12 gal Bags; 1/Pk
Representative image only.
Clavies Biohazard Bag Stand with Tray for 10 to 12 gal Bags; 1/Pk
| Item: 0649717
Securely supports biohazard bags and contents on benchtop or floor
₹34,000.00INR / Each
₹34,000.00INR / Each
Product Description
Key Features
  • Stands ship flat and are easy to assemble
  • Polypropylene bottom serves as containment tray
More About this Item
Clavies stainless steel and polypropylene bag stands are designed to securely support biohazard bags and their contents. Tray is safety orange for easy identification as a biohazard containment system. Stands are lightweight and easy to assemble. Autoclavable
Fits bags: 24 x 30" (61 x 76 cm)
Specifications
  • Capacity (Gal)12
  • ColorOrange and silver
  • DescriptionClavies Biohazard Bag Stand with Tray for 10 to 12 gal Bags; 1/Pk