Chrome-Plated Steel Jar Sleeve for Milling Jar 04172-00 (0.125 gal)
Representative image only.
Chrome-Plated Steel Jar Sleeve for Milling Jar 04172-00 (0.125 gal)
| Mfr: ST-0016| Item: 0417210
₹121,000.00INR / Each
₹121,000.00INR / Each
Product Description
Key Features
  • 3 month warranty
More About this Item
Jar sleeves increase rigidity and traction. Order HDPE milling jars separately.
3 Month Warranty
Specifications
  • Jar Capacity (Gal)0.125
  • MaterialStainless Steel
  • DescriptionChrome-Plated Steel Jar Sleeve for Milling Jar 04172-00 (0.125 gal)
  • Warranty3 Months