Chem Express X593 Mercury Vapor Badge with PrePaid Analysis Pk 5
Representative image only.
Chem Express X593 Mercury Vapor Badge with PrePaid Analysis Pk 5
Chem Express | Mfr: X593| Item: 8637834
Analysis performed in an AIHA-accredited lab
₹103,000.00INR / Pkg of 5
₹103,000.00INR / Pkg of 5
Product Description
Key Features
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
More About this Item
ChemExpress™ Mercury Vapor Badge monitors mercury vapor commonly emitted from mining operations and fluorescent lamps. Mercury vapor is collected by a thin ultra high-purity layer of gold. Monitors low levels of mercury with a detection limit of 0.0002 ppm over an 87 hour period. Each badge comes with a particle screen to prevent particles containing mercury from entering the sampler during monitoring and causing a high biased result. An analysis of elemental mercury is performed by cold vapor atomic absorption (CVAA).
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. B
1 Year Warranty
Specifications
  • Gases MeasuredMercury
  • DescriptionMercury Vapor Badge with PrePaid Analysis Pk 5