Chem Express X586 Ozone Badge with PrePaid Analysis Pk 5
Representative image only.
Chem Express X586 Ozone Badge with PrePaid Analysis Pk 5
Chem Express | Mfr: X586| Item: 8637833
Analysis performed in an AIHA-accredited lab
₹79,100.00INR / Pkg of 1
₹79,100.00INR / Pkg of 1
Product Description
Key Features
  • Save money and time with prepaid analysis
  • Lightweight and easy to use
  • 1 year warranty
More About this Item
ChemExpress™ Ozone Monitor Badge is a sensitive badge which allows sampling for Ozone commonly emitted by UV lamps, printers and copiers. Ozone is collected indirectly on fiberglass treated with nitrite. The filter ids then extracted with deionized water and analyzed by ion chromatography. A low 8 hour detection limit of 0.008ppm is made possible by using a ultra pure source of nitrite.
Personal monitoring badges are the most cost effective and efficient ways to monitor chemicals in your environment. Badges are easy to use and require minimal training and experience. They are small and light weight and have minimal effect on worker productivity. Badges are diffusive devices that require no additional equipment such as sampling pumps, calibration materials, or chargers. These long
1 Year Warranty
Specifications
  • Gases MeasuredOzone
  • DescriptionOzone Badge with PrePaid Analysis Pk 5