CELLTREAT Scientific Products 229870 Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs
Representative image only.
CELLTREAT Scientific Products 229870 Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs
CELLTREAT Scientific Products | Mfr: 229870| Item: 0193180
Get improved gas exchange via oversized vent area
₹76,100.00INR / Case of 4
₹76,100.00INR / Case of 4
Buy It Together
Current Item
CELLTREAT Scientific Products 229870 Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs
CELLTREAT Scientific Products 229870 Vented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs
0193180
₹76,100.00INR / Case of 4
+ =
Total Price 124100.00 ADD 2 ITEMS TO CART
Product Description
Includes
Vented sterile HDPE caps.
Key Features
  • Fernbach style offers a large surface area to volume ratio
  • Fernbach flasks with plain bottom
  • Fernbach flask shape increases surface area to volume ratio
More About this Item
Designed for suspension culture applications, these flasks feature 70-mm opening for easy filling and dispensing. Constructed of optically clear, autoclavable polycarbonate. All flasks are supplied sterile (gamma irradiated) and individually double bagged.
Sterile
Specifications
  • MaterialPolycarbonate
  • Capacity (mL)4000
  • AutoclavableYes
  • Working Volume (mL)3000
  • DescriptionVented Sterile Fernbach Flask with Plain Bottom, 3 L, Individually Wrapped; 4/cs