CELLTREAT Scientific Products 229865 Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
Representative image only.
CELLTREAT Scientific Products 229865 Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
CELLTREAT Scientific Products | Mfr: 229865| Item: 0193179
Get improved gas exchange via oversized vent area
₹83,600.00INR / Case of 6
₹83,600.00INR / Case of 6
Buy It Together
Current Item
CELLTREAT Scientific Products 229865 Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
CELLTREAT Scientific Products 229865 Vented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
0193179
₹83,600.00INR / Case of 6
+ =
Total Price 131600.00 ADD 2 ITEMS TO CART
Product Description
Includes
Vented sterile HDPE caps.
Key Features
  • Fernbach style offers a large surface area to volume ratio
  • Fernbach flasks with baffled bottom
  • Fernbach flask shape increases surface area to volume ratio
  • Baffled bottoms aid agitation and aeration during shaking
More About this Item
Designed for suspension culture applications, these flasks feature 70-mm opening for easy filling and dispensing. Constructed of optically clear, autoclavable polycarbonate. All flasks are supplied sterile (gamma irradiated) and individually double bagged.
Sterile
Specifications
  • MaterialPolycarbonate
  • Capacity (mL)2400
  • AutoclavableYes
  • Working Volume (mL)2000
  • DescriptionVented Sterile Fernbach Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs