CELLTREAT Scientific Products 229855 Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
Representative image only.
CELLTREAT Scientific Products 229855 Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
CELLTREAT Scientific Products | Mfr: 229855| Item: 0193177
Get improved gas exchange via oversized vent area
₹79,800.00INR / Case of 6
₹79,800.00INR / Case of 6
Buy It Together
Current Item
CELLTREAT Scientific Products 229855 Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
CELLTREAT Scientific Products 229855 Vented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs
0193177
₹79,800.00INR / Case of 6
+ =
Total Price 127800.00 ADD 2 ITEMS TO CART
Product Description
Includes
Vented sterile HDPE caps.
Key Features
  • Fernbach style offers a large surface area to volume ratio
  • Erlenmeyer flask with baffled bottom
  • Baffled bottoms aid agitation and aeration during shaking
More About this Item
Designed for suspension culture applications, these flasks feature 70-mm opening for easy filling and dispensing. Constructed of optically clear, autoclavable polycarbonate. All flasks are supplied sterile (gamma irradiated) and individually double bagged.
Sterile
Specifications
  • MaterialPolycarbonate
  • Capacity (mL)2400
  • AutoclavableYes
  • Working Volume (mL)2000
  • DescriptionVented Sterile Erlenmeyer Flasks with Baffled Bottom, 2 L, Individually Wrapped; 6/cs