CELLTREAT Scientific Products 229785 Solution Bottles, 1000mL, Sterile; 24/CS
Representative image only.
CELLTREAT Scientific Products 229785 Solution Bottles, 1000mL, Sterile; 24/CS
CELLTREAT Scientific Products | Mfr: 229785| Item: 0633679
Perfectly pair with bottletop filtration units
₹89,700.00INR / Case of 24
₹89,700.00INR / Case of 24
Product Description
Includes
Cap.
Key Features
  • Designed with an indented grip area for easy handling
  • Compatible with CELLTREAT Scientific Products bottletop filters
  • Bottles can also be used as solution or storage containers
More About this Item
A polyethylene cap provides an airtight seal, minimizing the risk of contamination.
Sterile
Specifications
  • Housing MaterialPolystyrene (PS)
  • Capacity (mL)1000